BSA blocking buffer, 5% in TBS, with 0,05% Tween-20
Supplier: Thermo Fisher Scientific
Certificates
About this item
Storage Temperature: −30 to −10 °C
Physical Form: Liquid
Specifications
- Storage temperature:-30°C to -10°C
- Physical form:Liquid
Specifications
Related Information
Frequently Bought Together