
3-(Trifluoromethyl)-5,6,7,8-tetrahydro[1,2,4]triazolo[4,3-a]pyrazine hydrochloride ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:762240-92-6
:
Formula:C6H8ClF3N4
MDL Number:MFCD09817638
:228.60 g/mol
:GHS07
Related Information
Frequently Bought Together