
4-(Trifluoromethoxy)phenyl isothiocyanate 97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:64285-95-6
:
Formula:C₈H₄F₃NOS
MDL Number:MFCD00042412
:219.19 g/mol
Boiling Pt:215…218 °C (lit.)
Density:1,35
Flash Pt:107 °C (225 °F)
ADR:8,II
:GHS05
Related Information
Frequently Bought Together