Palladium ≥99.95% (metals basis), powder , APS 0.35-0.8 micron
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:7440-05-3
:
Formula:Pd
MDL Number:MFCD00011167
:106.420 g/mol
Storage Temperature:Ambient
EINECS:231-115-6
UN:3089
ADR:4.1,II
:GHS02
Specifications
- Environmentally Preferable:
Specifications
Related Information
Frequently Bought Together