Order Entry
Ireland
ContactUsLinkComponent
4-Hydroxyindanone 97%
4-Hydroxyindanone 97%
Catalog # H56184.06
CAS Number:  
4-Hydroxyindanone 97%
Catalog # H56184.06
CAS Number:  

Specifications

  • Pk:
    5 g
  • Seq:
    3173

Specifications

About this item

CAS Number:40731-98-4
 :
Formula:C₉H₈O₂
MDL Number:MFCD00143330
 :148.161 g/mol
Melting Pt:242…244 °C
 :GHS07
Storage Temperature:Ambient