N-Butyrophenone-p-toluenesulphonylhydrazone 97%
Catalog # H53428.06
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pk:5 g
- Seq:3173
Specifications
About this item
:
:316.42 g/mol
Melting Pt:121…125 °C
Storage Temperature:Ambient
CAS Number:41780-81-8
Formula:C₁₇H₂₀N₂O₂S
MDL Number:MFCD01230699