

cis-N-Boc-4-cyano-L-proline methyl ester 97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:487048-28-2
:
Formula:C₁₂H₁₈N₂O₄
MDL Number:MFCD03787928
:254.286 g/mol
:GHS06
:GHS07
Storage Temperature:Ambient
Related Information
Frequently Bought Together