

4-Amino-1-(3-chloro-2-pyridyl)piperidine hydrochloride 97%
Catalog # H51927.MD
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pk:250 mg
- Seq:2590
Specifications
About this item
CAS Number:77145-35-8
:
Formula:C₁₀H₁₄ClN₃·HCl
MDL Number:MFCD16251586
Melting Pt:265…267 °C
:248.15 g/mol
:GHS06
:GHS07
Storage Temperature:Ambient