Order Entry
Ireland
ContactUsLinkComponent
1-Oxo-3-phenyl-1-(1-piperidinyl)-(2S)-propylamine
1-Oxo-3-phenyl-1-(1-piperidinyl)-(2S)-propylamine
Catalog # H51004.03
CAS Number:  
1-Oxo-3-phenyl-1-(1-piperidinyl)-(2S)-propylamine
Catalog # H51004.03
CAS Number:  

Specifications

  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

CAS Number:29618-19-7
 :
Formula:C₁₄H₂₀N₂O
MDL Number:MFCD01475890