
2-Amino-4,5,6,7-tetrahydrobenzo[b]thiophene-3-carboxylic acid amide 96%
Catalog # H34245.06
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pk:5 g
- Seq:3173
Specifications
About this item
CAS Number:4815-28-5
:
Formula:C₉H₁₂N₂OS
MDL Number:MFCD00111277
:196.27 g/mol
Melting Pt:195…197 °C
:GHS07
Storage Temperature:Ambient