3-Bromocatechol 95%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:14381-51-2
:
Formula:C₆H₅BrO₂
MDL Number:MFCD00869768
:189.01 g/mol
Melting Pt:39…42 °C
:GHS07
Storage Temperature:Ambient
Related Information
Frequently Bought Together