
1-Pyridin-3-ylethylamine 96%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:56129-55-6
:
Formula:C₇H₁₀N₂
MDL Number:MFCD01691641
:122.17 g/mol
ADR:8,III
:GHS05
Storage Temperature:Ambient
Related Information
Frequently Bought Together