

2-Hydroxy-2-(4-methylphenyl)acetic acid ≥98%, ee 97%, ChiPros®
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:31284-89-6
:MFCD00798433
Formula:C₉H₁₀O₃
MDL Number:MFCD00961860
:166.18 g/mol
Melting Pt:130…133 °C
:GHS05
:GHS07
Storage Temperature:Ambient
Related Information
Frequently Bought Together