5-Norbornene-2-carboxaldehyde (endo + exo mixture) 95%
: Bicyclo[2.2.1]hept-5-ene-2-carboxaldehyde
:5453-80-5
:
:C₈H₁₀O
:MFCD00167568
:122.17 g/mol
:68…70 °C (12 mmHg)
:1,032
:50 °C (122 °F)
:Ambient
:226-698-9
:1989
:3,III
:GHS02
- Environmentally Preferable:
Frequently Bought Together