6-Hydroxyhexanoic acid 95% may cont. variable amounts of dimer
Supplier: Thermo Fisher Scientific
Certificates
About this item
Synonyms: 6-Hydroxycaproic acid
CAS Number:1191-25-9
:
Formula:C₆H₁₂O₃
MDL Number:MFCD00046560
Density:1.08
Molecular Weight:132.159 g/mol
Flash Pt:>110 °C (230 °F)
:GHS07
Specifications
- Environmentally Preferable:
Specifications
Related Information
Frequently Bought Together