4-MUG 98%
Supplier: Thermo Fisher Scientific
Jump to:
About this item
Purity: 98% CAS No.: 6160-80-1 Molecular Formula: C16H16O9 Formula Weight: 352.30 Storage Temperature: -30°C to -10°C Optical Rotation: -106° (c=0.1 in water) MDL No.: MFCD00036772
Synonyms: (2S,3S,4S,5R,6S)-3,4,5-Trihydroxy-6-((4-methyl-2-oxo-2H-chromen-7-yl)oxy)tetrahydro-2H-pyran-2-carboxylic acid, 4-methylumbelliferyl-beta-D-glucuronide, 4-Methylumbelliferyl-β-D-glucuronide
- CAS Number:6160-80-1
- Formula:C₁₆H₁₆O₉
- MDL Number:MFCD00150472
- Molecular Weight:352.3 g/mol
- Storage Temperature:Freezer
- EINECS:228-186-0
2 Options Available Below
Product Details & Documents
Purity: 98% CAS No.: 6160-80-1 Molecular Formula: C16H16O9 Formula Weight: 352.30 Storage Temperature: -30°C to -10°C Optical Rotation: -106° (c=0.1 in water) MDL No.: MFCD00036772
Synonyms: (2S,3S,4S,5R,6S)-3,4,5-Trihydroxy-6-((4-methyl-2-oxo-2H-chromen-7-yl)oxy)tetrahydro-2H-pyran-2-carboxylic acid, 4-methylumbelliferyl-beta-D-glucuronide, 4-Methylumbelliferyl-β-D-glucuronide
- CAS Number:6160-80-1
- Formula:C₁₆H₁₆O₉
- MDL Number:MFCD00150472
- Molecular Weight:352.3 g/mol
- Storage Temperature:Freezer
- EINECS:228-186-0



