Carbon disulphide ≥99.9% ACS
Catalog # 39785.AY
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pk:4
- Seq:0004
- Environmentally Preferable:
Specifications
About this item
:
:76.14 g/mol
Merck Index:14,1811
Boiling Pt:46 °C
Melting Pt:–112 °C
Density:1,263
Flash Pt:–30 °C (–22 °F)
EINECS:200-843-6
UN:1131
ADR:3,I
:GHS02
:GHS08
:GHS07
Storage Temperature:Ambient
CAS Number:75-15-0
Formula:CS₂
MDL Number:MFCD00011321