

4-Oxo-4-phenyl-2-butenoic acid predominantly trans 97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:583-06-2
:
Formula:C₁₀H₈O₃
MDL Number:MFCD00014015
:176.17 g/mol
Melting Pt:93…97 °C
Storage Temperature:Ambient
EINECS:209-496-5
UN:2811
ADR:6.1,III
:GHS06
:GHS07
Related Information
Frequently Bought Together