Order Entry
Ireland
ContactUsLinkComponent
 
Bismuth ≥99.9999% (metals basis), rods, Puratronic®, Length: 145 mm (5.71 in), Ø 10 mm (0.39 in)
Catalog # 47229.KF
CAS Number:  
Bismuth ≥99.9999% (metals basis), rods, Puratronic®, Length: 145 mm (5.71 in), Ø 10 mm (0.39 in)
Catalog # 47229.KF
CAS Number:  

Specifications

  • Pk:
    1
  • Seq:
    0001
  • Environmentally Preferable:

Specifications

About this item

CAS Number:7440-69-9
 :
Formula:Bi
MDL Number:MFCD00134033
 :208.98 g/mol
Storage Temperature:Ambient
EINECS:231-177-4