Methoxypolyethylene glycol amine, M.W. 13,000
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:80506-64-5
:
Formula:H₂NCH₂CH₂(OCH₂CH₂)nOCH₃
MDL Number:MFCD00131599
Storage Temperature:Ambient
Related Information
Frequently Bought Together