Copper 65, 65 Cu, plasma standard solution, 65 Cu 10 µg/ml, Specpure®
45276.AD
:
- Concentration:10 mg/L
- Matrix:2% HNO₃
- Pk:50 mL
- MDL No.:MFCD00144006
- Element:Copper
- Symbol:Cu
Molecular Formula: Matrix: 2% HNO₃
Physical Form: Liquid
UN No.: UN3264
MDL No.: MFCD00144006