Titanium(IV) oxide 20 - 35% in water, colloidal dispersion
Catalog # 44517.A1
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pk:1 kg
- Seq:4024
- Environmentally Preferable:
Specifications
About this item
Synonyms: Titanic anhydride, Pigment White 6, Rutile, Titanium(IV) oxide, rutile, Titanium dioxide
CAS Number:13463-67-7
:
Formula:TiO₂
MDL Number:MFCD00011269
:79.88 g/mol
:GHS08
Storage Temperature:Ambient
:1317-70-0
Boiling Pt:368 °C
Melting Pt:1855
Density:1.26
Merck Index:13,09549