Order Entry
Ireland
ContactUsLinkComponent
Nickel (II) nitrate 1% Ni(NO3)2 in nitric acid 2%, Specpure®
Health
Corrosive
Nickel (II) nitrate 1% Ni(NO3)2 in nitric acid 2%, Specpure®
Catalog # 39043.AE
CAS Number:  
Health
Corrosive
Nickel (II) nitrate 1% Ni(NO3)2 in nitric acid 2%, Specpure®
Catalog # 39043.AE
CAS Number:  

Specifications

  • Pk:
    100 mL
  • Seq:
    2440

Specifications

About this item

 :
 :182.7 g/mol
Storage Temperature:Ambient
UN:3264
ADR:8,III
 :GHS08
 :GHS05
CAS Number:13138-45-9
Formula:N₂NiO₆
MDL Number:MFCD00011139