Cerium(III) ammonium nitrate tetrahydrate, Reagent Grade
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:13083-04-0
:
Formula:H₈CeN₇O₁₅·4H₂O
MDL Number:MFCD00210608
Molecular Weight:558.28 g/mol (486.22 anhy)
UN:1477
ADR:5.1 III
:GHS03
:GHS07
Storage Temperature:Ambient
Specifications
- Environmentally Preferable:
Specifications
Related Information
Frequently Bought Together