Order Entry
Ireland
Orders LinkContactUsLinkComponent
Boron phosphate

Boron phosphate

Boron phosphate
Certificates
Boron phosphate

2 Options Available Below

Product Details & Documents

CAS No.: 13308-51-5 Molecular Formula: BPO4 Formula Weight: 105.79 Physical Form: Powder Density: 2.81 Appearance: White Odour: Odourless Melting Point: 1400° MDL No.: MFCD00011318

  • CAS Number:13308-51-5
  • Formula:BO₄P
  • MDL Number:MFCD00011318
  • Molecular Weight:105.78 g/mol
  • Melting Pt:>1400 °C
  • Density:2,81
  • Storage Temperature:Ambient
  • EINECS:236-337-7
Product Family Options

Product Information