


Zirconium(IV) Propoxide ≥70% (w/w) in n-propanol
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:23519-77-9
:
Formula:C₁₂H₂₈O₄Zr
MDL Number:MFCD00015307
:327.58 g/mol
Density:1,044
Flash Pt:28 °C (83 °F)
EINECS:245-711-9
UN:1993
ADR:3,III
:GHS05
:GHS02
:GHS07
Storage Temperature:Ambient
Related Information
Frequently Bought Together