Order Entry
Ireland
ContactUsLinkComponent
Cover glass for haemocytometers
  HAWKZ50000
Cover glass for haemocytometers
  HAWKZ50000
  • L×W:
    22×25 mm
  • Thickness:
    0,5 mm
  • Pk:
    10
Cover glass with polished surfaces for accurate observations, in laboratory and hospital environments.