Order Entry
Ireland
ContactUsLinkComponent
Horizontal electrophoresis system, multiSUB midi
Horizontal electrophoresis system, multiSUB midi
Supplier:  CLEAVER SCIENTIFIC
Certificates
undefined
Horizontal electrophoresis system, multiSUB midi
Supplier:  CLEAVER SCIENTIFIC
Specifications
  • Catalog No:
  • CLEVMSMIDI7
  • CLEVMSMIDI10
  • CLEVMSMIDIDUO
  • Model:
  • multiSUB midi 7
  • multiSUB midi 10
  • multiSUB midi DUO
  • Gel size W×L:
  • 100×70 mm
  • 100×100 mm
  • 100×70 ; 100×100 mm
  • Sample capacity:
  • 50 samples
  • 100 samples
  • Buffer volume:
  • 300 ml
  • 300 ml
  • 300 ml
  • Unit W×L×H:
  • 125×220×90 mm
  • 125×220×90 mm
  • 125×220×90 mm
Specifications
Combs
Product Information
Frequently Bought Together