Order Entry
Ireland
ContactUsLinkComponent
Anti-ULK1 Rabbit Polyclonal Antibody
Anti-ULK1 Rabbit Polyclonal Antibody
Catalog # ABGEAP19250C
Supplier:  Abgent
Anti-ULK1 Rabbit Polyclonal Antibody
Catalog # ABGEAP19250C
Supplier:  Abgent

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Unc-51 like autophagy activating kinase 1
  • Antigen symbol:
    ULK1
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Epitope:
    internal
  • Gene ID:
    8408
  • Antigen synonyms:
    ATG1|ATG1A|hATG1|Unc51.1|UNC51
  • Amino acid number:
    296 to 322
  • Storage buffer:
    pH 7,4
  • Molecular weight:
    112.631 kDa
  • Sequence:
    HLASPPSLGEMQC
  • Storage temperature:
    Maintain refrigerated at 2-8 °C for up to 6 months. For long term storage store at –20 °C in small aliquots to prevent freeze-thaw cycles
  • Concentration:
    0.36
  • Shipping temperature:
    4 °C
  • Immunogen:
    This ULK1 antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide between 296-322 amino acids from the Central region of human ULK1.
  • Purification:
    Protein A & Peptide affinity purification
  • Pk:
    400 µl

Specifications

About this item

Type: Primary
Antigen: ULK1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: internal
Host: Rabbit
Isotype:
Reactivity: Human