Anti-ASB3 Rabbit Polyclonal Antibody
ABGEAP16752A
: Abgent
- Antibody type:Primary
- Antigen symbol:ASB3
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human,Mouse
- Western blot:Yes
- Epitope:N-terminal
- Gene ID:100302652;51130
- Amino acid number:15 to 42
- Storage buffer:pH 7,4
- Molecular weight:57.745 kDa
- Sequence:LKKGRSVDVADNRC
- Storage temperature:Maintain refrigerated at 2-8 °C for up to 6 months. For long term storage store at –20 °C in small aliquots to prevent freeze-thaw cycles
- Concentration:0.55
- Shipping temperature:4 °C
- Immunogen:This ASB3 antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide between 15-42 amino acids from the N-terminal region of human ASB3.
- Purification:Protein A & Peptide affinity purification
- Pk:400 µl
Type: Primary
Antigen: ASB3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype:
Reactivity: Human, Mouse