Order Entry
Ireland
ContactUsLinkComponent
 
Anti-MTL5 Rabbit Polyclonal Antibody
  ABGEAP18102B
 :  Abgent
Anti-MTL5 Rabbit Polyclonal Antibody
  ABGEAP18102B
 :  Abgent

 

  • Antibody type:
    Primary
  • Antigen name:
    Metallothionein-like 5, testis-specific (tesmin)
  • Antigen symbol:
    MTL5
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Epitope:
    C-terminal
  • Gene ID:
    9633
  • Antigen synonyms:
    MTLT|TESMIN|CXCDC2
  • Amino acid number:
    341 to 368
  • Storage buffer:
    pH 7,4
  • Molecular weight:
    55.025 kDa
  • Sequence:
    KGQLGNVKPQHNKC
  • Storage temperature:
    Maintain refrigerated at 2-8 °C for up to 6 months. For long term storage store at –20 °C in small aliquots to prevent freeze-thaw cycles
  • Concentration:
    0.65
  • Shipping temperature:
    4 °C
  • Immunogen:
    This MTL5 antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide between 341-368 amino acids from the C-terminal region of human MTL5.
  • Purification:
    Protein A & Peptide affinity purification
  • Pk:
    400 µl

 

 

Type: Primary
Antigen: MTL5
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype:
Reactivity: Human