Order Entry
Ireland
ContactUsLinkComponent
 
Anti-ANP32E Rabbit Polyclonal Antibody
  ABGEAP20559A
 :  Abgent
undefined
Anti-ANP32E Rabbit Polyclonal Antibody
  ABGEAP20559A
 :  Abgent

 

  • Antibody type:
    Primary
  • Antigen name:
    Acidic (leucine-rich) nuclear phosphoprotein 32 family, member E
  • Antigen symbol:
    ANP32E
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • Reactivity:
    Human,
    Mouse
  • Western blot:
    Yes
  • Epitope:
    N-terminal
  • Gene ID:
    81611
  • Antigen synonyms:
    LANPL|LANP-L
  • Storage buffer:
    pH 7,4
  • Molecular weight:
    30.692 kDa
  • Sequence:
    NGEIEGLNDTFKEL
  • Storage temperature:
    Maintain refrigerated at 2-8 °C for up to 6 months. For long term storage store at –20 °C in small aliquots to prevent freeze-thaw cycles
  • Concentration:
    0.82
  • Shipping temperature:
    4 °C
  • Immunogen:
    This ANP32E antibody is generated from a rabbit immunized with a KLH conjugated synthetic peptide between 21-44 amino acids from the N-terminal region of human ANP32E.
  • Purification:
    Protein A & Peptide affinity purification
  • Pk:
    400 µl

 

 

Type: Primary
Antigen: ANP32E
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype:
Reactivity: Human, Mouse