Order Entry
Ireland
ContactUsLinkComponent
VWR® Barcelona, Safety Spectacles
VWR® Barcelona, Safety Spectacles
Catalog # 111-1848
Supplier:  VWR Collection
undefined
VWR® Barcelona, Safety Spectacles
Catalog # 111-1848
Supplier:  VWR Collection
Specifications
  • Type:
    Anti-UV (2-1,2), anti-scratch, anti-fog
  • Colour:
    Grey/blue
  • Lenses:
    Clear PC
  • Pk:
    1
Specifications
About this item
Modern and stylish design glasses with sturdy polycarbonate frame.
  • Nose pad made from soft material for extra comfort and to avoid slippage
  • Soft temple dual-injected rubber to fit different head profiles
  • Pantoscopic adjustment
Certifications: Compliant with EN 166, EN 170; VWR® EN 166 FT CE (frame); VWR® 2C-1,2 1 FT (lens)