Order Entry
Ireland
ContactUsLinkComponent
L(+)-Ascorbic acid 99.0-100.5% USP, Macron Fine Chemicals™
L(+)-Ascorbic acid 99.0-100.5% USP, Macron Fine Chemicals™
Catalog # 1852-10
CAS Number:  
undefined
L(+)-Ascorbic acid 99.0-100.5% USP, Macron Fine Chemicals™
Catalog # 1852-10
CAS Number:  

Specifications

  • Pack type:
    Glass bottle, amber, wide mouth
  • Pk:
    500 g
  • Seq:
    3645

Specifications

Specification Test Results

  • Meets U.S.P Requirements:
  • GMP Manufactured Product:
  • CAUTION: For Manufacturing, processing or repackaging:
  • Bulk Pharmaceutical Chemical:
  • USP - Appearance (fine, white to slightly yellow crystals):
    Passes Test
  • USP - Assay (C₆H₈O₆):
    99.0 - 100.5 %
  • USP - Specific Rotation [Å]²⁵^D (+):
    20.5 - 21.5 Degree
  • USP - Residue on Ignition:
    ≤0.1 %
  • USP - Identification A:
    Passes Test
  • USP - Identification B:
    Passes Test

Specification Test Results

About this item

 :
 :176.13 g/mol
Merck Index:13,00837
Boiling Pt:553 °C (1013 hPa)
Melting Pt:190…192 °C
Density:1,694 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:200-066-2
REACH:exempt
CAS Number:50-81-7
Formula:C₆H₈O₆
MDL Number:MFCD00064328