Order Entry
Ireland
ContactUsLinkComponent
Sodium dihydrogen phosphate dihydrate 98.0-102.0%, BAKER ANALYZED®, J.T.Baker®
Sodium dihydrogen phosphate dihydrate 98.0-102.0%, BAKER ANALYZED®, J.T.Baker®
Catalog # 3819-01
Supplier:  Avantor
CAS Number:  
undefined
Sodium dihydrogen phosphate dihydrate 98.0-102.0%, BAKER ANALYZED®, J.T.Baker®
Catalog # 3819-01
Supplier:  Avantor
CAS Number:  

Specifications

  • Pack type:
    Poly bottle, white, blue T/E cap
  • Pk:
    500 g
  • Seq:
    3645

Specifications

Specification Test Results

  • For Laboratory,Research,or Manufacturing Use:
    98.0 - 102.0%
  • Assay (NaH₂PO₄·2H₂O):
    98.0 - 102.0 %
  • pH of 5% Solution at 25°C:
    4.1 - 4.5
  • Chloride (Cl):
    ≤5 ppm
  • Insoluble Matter, Calcium and NH₄OH Precipitate:
    ≤0.01 %
  • Nitrogen Compounds (as N):
    ≤0.001 %
  • ACS - Sulfate (SO₄):
    ≤0.003 %
  • Arsenic (As):
    ≤0.5 ppm
  • Heavy Metals (as Pb):
    ≤0.001 %
  • Trace Impurities - Iron (Fe):
    ≤0.001 %

Specification Test Results

About this item

 :
 :156.01 g/mol
Merck Index:12,08806
Boiling Pt:? 650 °C (1013 hPa)
Melting Pt:60 °C
Density:1,92 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-449-2
REACH:01-2119489796-13
CAS Number:13472-35-0
Formula:NaH₂PO₄·2H₂O
MDL Number:MFCD00149209