Measuring cylinders, SP Wilmad-LabGlass
Supplier: LAB GLASS WILMAD
Certificates
About this item
Glass, tall form, Class B.
- Calibrated to deliver at 20 °C (TD)
- With reinforced bead and pouring lip
- Suitable for general laboratory use
Certifications: Measuring cylinders complies with ASTM E1272.
Frequently Bought Together