Flasks, round bottom, with standard ground joints, SP Wilmad-LabGlass
Catalog # LABGLG-7220-150
Supplier: LAB GLASS WILMAD
Specifications
- Capacity:300 ml
- Seq:02
- Pk:1
- Centre neck:24/40 NS
Specifications
About this item
Borosilicate glass 3.3, with short neck.
- Excellent chemical resistance
- High temperature resistance
- Minimal thermal expansion giving relatively high resistance to temperature changes