Order Entry
Ireland
ContactUsLinkComponent
Cut resistant gloves, uvex unidur 6659 foam
Cut resistant gloves, uvex unidur 6659 foam
Supplier:  UVEX
Certificates
undefined
Cut resistant gloves, uvex unidur 6659 foam
Supplier:  UVEX
Specifications
  • Catalog No:
  • 112-4446
  • 112-4445
  • 112-4448
  • 112-4447
  • 112-4444
  • Model:
  • 6659 foam
  • 6659 foam
  • 6659 foam
  • 6659 foam
  • 6659 foam
  • Ambidextrous / hand-specific:
  • Hand-specific
  • Hand-specific
  • Hand-specific
  • Hand-specific
  • Hand-specific
  • Disposable / reusable:
  • Reusable
  • Reusable
  • Reusable
  • Reusable
  • Reusable
  • Liner material:
  • HPPE
  • HPPE
  • HPPE
  • HPPE
  • HPPE
  • Finish:
  • Nitrile Butadiene Rubber (NBR) foam coating on palm and fingertips
  • Nitrile Butadiene Rubber (NBR) foam coating on palm and fingertips
  • Nitrile Butadiene Rubber (NBR) foam coating on palm and fingertips
  • Nitrile Butadiene Rubber (NBR) foam coating on palm and fingertips
  • Nitrile Butadiene Rubber (NBR) foam coating on palm and fingertips
  • Cuff style:
  • Knit wrist
  • Knit wrist
  • Knit wrist
  • Knit wrist
  • Knit wrist
  • EN norm:
  • EN 388:2016 (4X43C)
  • EN 388:2016 (4X43C)
  • EN 388:2016 (4X43C)
  • EN 388:2016 (4X43C)
  • EN 388:2016 (4X43C)