VWR®, Lens Cleaning Tissues
Catalog # VWRU52845-009
Supplier: VWR Collection
Specifications
- Packed:12×50 sheets
- Description:Lens cleaning tissue
- L×W:152×203 mm
- Seq:02
- Pk:12
Specifications
About this item
Suitable for cleaning safety glasses and goggles, microscope lenses and more.
- High wet strength
- Dust free lens
- No scratching