Order Entry
Ireland
ContactUsLinkComponent
VWR®TraceClean®, Water sampling bottles, amber glass
VWR®TraceClean®, Water sampling bottles, amber glass
Catalog # VWRU89094-064
Supplier:  VWR Collection
undefined
VWR®TraceClean®, Water sampling bottles, amber glass
Catalog # VWRU89094-064
Supplier:  VWR Collection
Specifications
  • Capacity:
    4000 ml
  • Description:
    Processed (PC)
  • Seq:
    03
  • Pk:
    4
Specifications
About this item
Type III (soda-lime) amber glass.
  • Available with solid-top caps with fluoropolymer resin liner are for use in semi-volatiles, pesticides, PCBs, and phenolics analyses
  • Amber glass is recommended for UV light-sensitive samples
Processed (PC) and Quality-Assured® (QA) packers follow EPA cleaning procedures listed in the OSWER Directive 9240.0-05A 'Specifications and Guidance for Contaminant-Free Sample Containers.' QA packers include a Certificate of Analysis in every case. QA packers also include bar codes with lot number and individual container number for complete traceability. Not Processed (NPC) packers do not receive the specialty cleaning process. All cases are custody-sealed to ensure content integrity.