Order Entry
Ireland
ContactUsLinkComponent
VWR® Talon®, Stand plate, rectangular
  VWRU12985-070
undefined
VWR® Talon®, Stand plate, rectangular
  VWRU12985-070
  • Description:
    Support stand base, rectangular
  • L×W:
    127×203 mm
  • Seq:
    02
  • Pk:
    1
Cast iron with black enamel finish.
  • All bases feature a built-in support rod holder with locking knob that accepts a 13 mm Ø support rod (sold separately)
  • For use with VWR® Talon® lab frames