Order Entry
Ireland
ContactUsLinkComponent
(±)-2-Methyltetrahydrofuran, EMPLURA®
Flammable
Corrosive
Irritant
(±)-2-Methyltetrahydrofuran, EMPLURA®
Supplier:  Merck
Certificates
Flammable
Corrosive
Irritant
(±)-2-Methyltetrahydrofuran, EMPLURA®
Supplier:  Merck
Certificates
Limited Time Sale
Select items below are currently on sale

Specification Test Results

  • Hill Formula:
    C₅H₁₀O
  • Molar Mass:
    86.13 g/mol
  • Boiling point:
    78 - 80 °C
  • Density:
    0.85 g/cm³ (20 °C)
  • Explosion limit:
    1.2 - 5.7 %(V)
  • Flash point:
    -12 °C
  • Ignition temperature:
    270 °C
  • Melting Point:
    -136 °C
  • Vapor pressure:
    130 hPa (25 °C)
  • Solubility:
    130 g/l (25 °C)
  • LD 50 oral:
    LD50 Rat300 - 2000 mg/kg
  • LD 50 dermal:
    LD50 Rat2000 mg/kg
  • Storage class:
    3 Flammable liquids
  • WGK:
    WGK 2 water endangering
  • R Phrase:
    R 11-19-22-38-41
  • S Phrase:
    S 16-29-33-26-39

Specification Test Results

Related Information

Frequently Bought Together