Anti-IgG Goat Antibody (HRP (Horseradish Peroxidase))
Catalog # 786-R41
Supplier: G-Biosciences
Specifications
- Antibody type:Secondary
- Antigen symbol:IgG
- Conjugation:HRP (Horseradish Peroxidase)
- ELISA:Yes
- Flow cytometry:Yes
- Host:Goat
- ImmunoChemistry:Yes
- Reactivity:Human
- Seq:04
- Pk:2 mL