Anti-IgG Goat Antibody (AP (Alkaline Phosphatase))
Catalog # 786-R43
Supplier: G-Biosciences
Specifications
- Antibody type:Secondary
- Antigen symbol:IgG
- Conjugation:AP (Alkaline Phosphatase)
- ELISA:Yes
- Flow cytometry:Yes
- Host:Goat
- ImmunoChemistry:Yes
- Reactivity:Mouse
- Seq:07
- Pk:1 mL