Anti-GST Goat Polyclonal Antibody (SureLight® R-Phycoerythrin)
Catalog # AGRIAS152914
Supplier: Agrisera
Specifications
- Antibody type:Primary
- Antigen name:Glutathione-S-Transferase
- Antigen symbol:GST
- Clonality:Polyclonal
- Conjugation:SureLight® RPE (R-Phycoerythrin)
- Flow cytometry:Yes
- Host:Goat
- Immunogen:Glutathione-S-transferase from S. japonicum
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Goat anti-GST is a primary antibody to GST directly conjugated to SureLight® R-Phycoerythrin.,Goat anti-GST is a primary antibody to GST directly conjugated to SureLight® R-Phycoerythrin.
Type: Primary
Antigen: GST
Clonality: Polyclonal
Clone:
Conjugation: SureLight® RPE (R-Phycoerythrin)
Epitope:
Host: Goat
Isotype:
Reactivity: