Specifications
- Antibody type:Primary
- Antigen symbol:PIF5
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Reactivity:A. thaliana
- Western blot:Yes
- Immunogen:KLH-conjugated synthetic peptide derived from Arabidopsis thaliana PIF5, UniProt: Q84LH8, TAIR: AT3G59060
- Size:50 µg
- Pk:50 µG
Specifications
About this item
PIF5 (Phytochrome interacting factor 5) is a transcription factor which acts negatively in the phytochrome B signaling pathway and regulates PHYB at post-transcriptional level. Alternative protein names: AtbHLH65, bHLH65, bHLH065, Basic helix-loop-helix protein 65, PHYTOCHROME INTERACTING FACTOR 3-LIKE 6, PIL6, Phytochrome interacting factor-like 6, Transcription factor EN 103.,PIF5 (Phytochrome interacting factor 5) is a transcription factor which acts negatively in the phytochrome B signaling pathway and regulates PHYB at post-transcriptional level. Alternative protein names: AtbHLH65, bHLH65, bHLH065, Basic helix-loop-helix protein 65, PHYTOCHROME INTERACTING FACTOR 3-LIKE 6, PIL6, Phytochrome interacting factor-like 6, Transcription factor EN 103.
Type: Primary
Antigen: PIF5
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: A. thaliana