VWR®, Polyester Films for ELISA and General Incubation
Catalog # VWRU60941-120
Supplier: VWR Collection
Specifications
- Sterile:-
- Description:Films
- Material:Polyester
- Thickness:50 µm
- Pk:100
Specifications
About this item
These 50 μm thick polyester general-purpose films are designed for ELISA, EIA, and incubation. Films feature a single end tab with no perforations and are suppled non sterile.
Each film L×D: 133×83 mm