VWR® Tubing, Rubber, Vacuum
Natural rubber, black/red.
- Flexible, very good tensile strength
- Seamless and acid-proof
- It also offers high resilience and tear resistance
Heavy wall, suitable for vacuum applications in the laboratory.
Hardness (Shore durometer A): 50
Tensile strength: 172 to 241 bar (black) 138 to 179 bar (red)
Frequently Bought Together