Accessories for reagent sprayer
Supplier: CAMAG
Certificates
About this item
Frequently Bought Together
Acetone ≥99%, TECHNICAL
Supplier: VWR Chemicals
Glass capillaries and graduated micropipettes, disposable
Supplier: CAMAG
VWR®, Snap Cap Vials
Supplier: VWR Collection
Benzethonium chloride 0.004 mol/l, AVS TITRINORM volumetric solution
Supplier: VWR Chemicals
VWR®, Screw Caps for Short Thread Vials ND9
Supplier: VWR Collection
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
Supplier: VWR Chemicals
VWR®, Crimp Neck Vials ND11
Supplier: VWR Collection
Pipette tips
Supplier: Sartorius
Water, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals
Bromophenol blue ACS, Reag. Ph. Eur., Supelco®
Supplier: Merck
Ammonia solution 32%, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
Test sieves, stainless steel
Supplier: ENDECOTTS
Dimidium bromide
Supplier: Apollo Scientific
Accessories for reagent sprayer
Supplier: CAMAG