Order Entry
Ireland
ContactUsLinkComponent
 
 
 
Tetra-tert-butyl orthotitanate for synthesis, Sigma-Aldrich®
  8.18320.0025
 :  Merck
 :  
 
 
Tetra-tert-butyl orthotitanate for synthesis, Sigma-Aldrich®
  8.18320.0025
 :  Merck
 :  

 

  • Pack type:
    Glass bottle
  • Pk:
    25 ml
  • Seq:
    2315

 

 

  • Solubility:
    (20 °C) (decomposition)
  • Molar Mass:
    340.32 g/mol
  • Boiling Point:
    160 - 165 °C (20 hPa)
  • Vapor Pressure:
    43 hPa (136 °C)
  • Refractive Index:
    1.4368 (20 °C, 589 nm)
  • Density:
    0.89 g/cm3 (20 °C)
  • Synonyms:
    tert-Butyl titanate(IV), Orthotitanic acid tetra-tert-butyl ester, Titanium tetra-tert-butylate, Titanic acid tetra-tert-butyl ester
  • Hill Formula:
    C16H36O4Ti
  • Chemical Formula:
    Ti(OC(CH3)3)4

 

 

 :
 :340.32 g/mol
 :3,III
 :M-Clarity™ quality level = MQ200
 :221-412-9
 :1993
 :GHS02
 :GHS07
 :Ambient
 :3087-39-6
 :C₁₆H₃₆O₄Ti
 :MFCD00040554